siRNA Details
Gene Symbol: Sppl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014200.1 | NP_001014222.1 | 5 | 474 | ||
XM_006240947.2 | XP_006241009.1 | 5 | 541 | |||
XM_006240948.2 | XP_006241010.1 | 5 | 541 | |||
XM_006240952.2 | XP_006241014.1 | 5 | 541 | |||
XM_008765108.1 | XP_008763330.1 | 4 | 979 | |||
XM_008765109.1 | XP_008763331.1 | 4 | 979 | |||
XM_008765110.1 | XP_008763332.1 | 4 | 979 | |||
XM_008765111.1 | XP_008763333.1 | 5 | 542 | |||
XM_008765112.1 | XP_008763334.1 | 5 | 542 | |||
XM_008765113.1 | XP_008763335.1 | 4 | 979 | |||
XM_008765114.1 | XP_008763336.1 | 4 | 979 | |||
XM_008765115.1 | XP_008763337.1 | 4 | 979 | |||
XM_008765116.1 | XP_008763338.1 | 4 | 979 | |||
XM_008765117.1 | XP_008763339.1 | 4 | 979 | |||
GenBank mRNA | BC087132.1 | - | 5 | 474 | 7121303 | |
FQ223857.1 | - | 6 | 624 |
