siRNA Details
Gene Symbol: Lsm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108732.1 | NP_001102202.1 | 2 | 79 | ||
XM_006240958.2 | XP_006241020.1 | 2 | 91 | |||
XM_008765118.1 | XP_008763340.1 | 2 | 81 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
