siRNA Details
Gene Symbol: Mbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108735.1 | NP_001102205.1 | 4 | 752 | ||
XM_006240965.1 | XP_006241027.1 | 4 | 746 | |||
XM_006240966.1 | XP_006241028.1 | 4 | 645 | |||
XM_008765119.1 | XP_008763341.1 | 5 | 624 | |||
XM_008765120.1 | XP_008763342.1 | 4 | 564 | |||
XM_008765121.1 | XP_008763343.1 | 5 | 548 | |||
XM_008765122.1 | XP_008763344.1 | 4 | 744 | |||
GenBank mRNA | BC167041.1 | - | 4 | 519 | ||
FQ216718.1 | - | 3 | 260 |
