siRNA Details
Gene Symbol: Mier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108737.1 | NP_001102207.1 | 9 | 896 | ||
XM_006240977.2 | XP_006241039.1 | 9 | 1131 | |||
XM_006240978.1 | XP_006241040.1 | 9 | 918 |
