siRNA Details
Gene Symbol: LOC100362745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763841.1 | XP_008762063.1 | 3 | 985 |
Gene Symbol: RGD1560314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765189.1 | XP_008763411.1 | 4 | 1783 | ||
GenBank mRNA | U78121.1 | - | 1 | 46 |
