siRNA Details
Gene Symbol: Cd320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014201.1 | NP_001014223.1 | 3 | 358 | ||
XM_006241128.2 | XP_006241190.1 | 3 | 2319 | |||
GenBank mRNA | BC088861.1 | - | 3 | 358 | 7310701 | |
FQ219198.1 | - | 3 | 360 | |||
FQ234194.1 | - | 3 | 371 |

More Information
Other Species Targeted

Gene Ontology Categories:
