siRNA Details
Gene Symbol: Ckap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108740.1 | NP_001102210.1 | 2 | 290 | ||
XM_008765245.1 | XP_008763467.1 | 2 | 923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
