siRNA Details
Gene Symbol: Appl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108741.1 | NP_001102211.1 | 19, 20 | 1897 | ||
XM_006241201.2 | XP_006241263.1 | 19, 20 | 1938 | |||
GenBank mRNA | BC168167.1 | - | 19, 20 | 1812 | 9021772 |
