siRNA Details
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007750.1 | NP_001007751.1 | 4, 5 | 748 | ||
XM_006241228.2 | XP_006241290.1 | 4, 5 | 743 | |||
XM_006241229.2 | XP_006241291.1 | 4, 5 | 743 | |||
XM_006241230.2 | XP_006241292.1 | 4, 5 | 743 | |||
XM_006241231.2 | XP_006241293.1 | 4, 5 | 743 | |||
GenBank mRNA | BC082074.1 | - | 4, 5 | 748 | 7133149 |

More Information
Other Species Targeted

Gene Ontology Categories:
