siRNA Details
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007750.1 | NP_001007751.1 | 2 | 386 | ||
XM_006241228.2 | XP_006241290.1 | 2 | 381 | |||
XM_006241229.2 | XP_006241291.1 | 2 | 381 | |||
XM_006241230.2 | XP_006241292.1 | 2 | 381 | |||
XM_006241231.2 | XP_006241293.1 | 2 | 381 | |||
GenBank mRNA | BC082074.1 | - | 2 | 386 | 7133149 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144807, NM_001146690, BY341208, BC031435, AK007964, and others.
Gene Ontology Categories:
