siRNA Details
Gene Symbol: Mybpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765246.1 | XP_008763468.1 | 30 | 3624 | ||
XM_008765247.1 | XP_008763469.1 | 30 | 3624 | |||
XM_008765248.1 | XP_008763470.1 | 29 | 3587 | |||
XM_008765249.1 | XP_008763471.1 | 29 | 3580 | |||
XM_008765250.1 | XP_008763472.1 | 29 | 3565 | |||
XM_008765251.1 | XP_008763473.1 | 28 | 3546 | |||
XM_008765252.1 | XP_008763474.1 | 28 | 3543 | |||
XM_008765253.1 | XP_008763475.1 | 29 | 3564 | |||
XM_008765254.1 | XP_008763476.1 | 28 | 3527 | |||
XM_008765255.1 | XP_008763477.1 | 28 | 3546 | |||
XM_008765256.1 | XP_008763478.1 | 27 | 3502 | |||
XM_008765257.1 | XP_008763479.1 | 27 | 3483 | |||
XM_008765258.1 | XP_008763480.1 | 27 | 3482 | |||
GenBank mRNA | FQ214958.1 | - | 5 | 677 | ||
FQ215501.1 | - | 6 | 768 | |||
X90475.1 | - | 15 | 1993 |
