siRNA Details
Gene Symbol: Mybpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765246.1 | XP_008763468.1 | 13 | 1261 | ||
XM_008765247.1 | XP_008763469.1 | 13 | 1261 | |||
XM_008765248.1 | XP_008763470.1 | 12 | 1224 | |||
XM_008765249.1 | XP_008763471.1 | 12 | 1217 | |||
XM_008765250.1 | XP_008763472.1 | 13 | 1256 | |||
XM_008765251.1 | XP_008763473.1 | 11 | 1183 | |||
XM_008765252.1 | XP_008763474.1 | 11 | 1180 | |||
XM_008765253.1 | XP_008763475.1 | 13 | 1255 | |||
XM_008765254.1 | XP_008763476.1 | 12 | 1218 | |||
XM_008765255.1 | XP_008763477.1 | 11 | 1183 | |||
XM_008765256.1 | XP_008763478.1 | 10 | 1139 | |||
XM_008765257.1 | XP_008763479.1 | 11 | 1174 | |||
XM_008765258.1 | XP_008763480.1 | 11 | 1173 |
