siRNA Details
Gene Symbol: R3hdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130557.1 | NP_001124029.1 | 8 | 750 | ||
XM_006241469.2 | XP_006241531.1 | 10 | 1835 | |||
XM_006241470.2 | XP_006241532.1 | 9 | 1691 | |||
XM_006241471.2 | XP_006241533.1 | 10 | 1801 | |||
XM_006241472.2 | XP_006241534.1 | 8 | 1599 | |||
XM_006241473.2 | XP_006241535.1 | 10 | 1740 | |||
XM_006241475.2 | XP_006241537.1 | 10 | 1835 | |||
XM_006241476.2 | XP_006241538.1 | 10 | 1835 | |||
XM_006241477.2 | XP_006241539.1 | 10 | 1835 | |||
XM_006241478.2 | XP_006241540.1 | 10 | 1835 | |||
XM_006241479.2 | XP_006241541.1 | 10 | 1835 | |||
XM_006241480.2 | XP_006241542.1 | 10 | 1835 | |||
XM_006241481.2 | XP_006241543.1 | 10 | 1834 | |||
XM_006241482.2 | XP_006241544.1 | 10 | 1834 | |||
XM_006241483.2 | XP_006241545.1 | 9 | 1690 | |||
XM_006241484.2 | XP_006241546.1 | 10 | 1834 | |||
XM_008765408.1 | XP_008763630.1 | 11 | 1146 | |||
XM_008765409.1 | XP_008763631.1 | 11 | 1247 | |||
XM_008765410.1 | XP_008763632.1 | 11 | 1014 | |||
XM_008765411.1 | XP_008763633.1 | 10 | 933 | |||
XM_008765412.1 | XP_008763634.1 | 10 | 1043 | |||
XM_008765413.1 | XP_008763635.1 | 9 | 947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
