siRNA Details
Gene Symbol: Nudcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130561.2 | NP_001124033.1 | 6 | 915 | ||
XM_006241614.2 | XP_006241676.1 | 6 | 1011 | |||
GenBank mRNA | BC169014.1 | - | 6 | 915 | 9091867 |
