siRNA Details
Gene Symbol: Mal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198786.2 | NP_942081.2 | 3 | 579 | ||
GenBank mRNA | AY079080.1 | - | 3 | 439 | ||
AY079081.2 | - | 2 | 692 | |||
BC062388.1 | - | 3 | 579 | 5620891 |
Gene Symbol: LOC100911027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748700.3 | XP_003748748.1 | 3 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
