siRNA Details
Gene Symbol: Derl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014202.1 | NP_001014224.1 | 3 | 369 | ||
GenBank mRNA | BC085877.1 | - | 3 | 369 | 7191715 | |
FQ218639.1 | - | 3 | 406 | |||
FQ220628.1 | - | 3 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
