siRNA Details
Gene Symbol: Derl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014202.1 | NP_001014224.1 | 7, 8 | 663 | ||
GenBank mRNA | BC085877.1 | - | 7, 8 | 663 | 7191715 | |
FQ218639.1 | - | 7, 8 | 700 | |||
FQ220628.1 | - | 7, 8 | 702 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024207, AK007435, AK161702, AK152197, AK043420, and others.
Gene Ontology Categories:
