siRNA Details
Gene Symbol: RGD1311188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135897.1 | NP_001129369.1 | 4 | 1137 | ||
XM_008765570.1 | XP_008763792.1 | 4 | 1108 | |||
GenBank mRNA | BC162017.1 | - | 4 | 1137 | 9098233 |
Gene Symbol: LOC103692939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765624.1 | XP_008763846.1 | 1 | 321 |
Gene Symbol: Zfp707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135903.1 | NP_001129375.1 | 4 | 1131 | ||
XM_006241815.2 | XP_006241877.1 | 4 | 1361 | |||
XM_006241816.2 | XP_006241878.1 | 4 | 1359 | |||
GenBank mRNA | BC161959.1 | - | 4 | 1131 | 9027463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
