siRNA Details
Gene Symbol: Fam83h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130565.1 | NP_001124037.1 | 5 | 3351 | ||
GenBank mRNA | FQ222515.1 | - | 4 | 2967 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam83h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130565.1 | NP_001124037.1 | 5 | 3351 | ||
GenBank mRNA | FQ222515.1 | - | 4 | 2967 |