siRNA Details
Gene Symbol: Nol12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012747.2 | NP_001012765.1 | 5 | 466 | ||
XM_006242007.2 | XP_006242069.1 | 5 | 742 | |||
GenBank mRNA | AY917132.1 | - | 5 | 444 | ||
BC103643.1 | - | 5 | 466 | 7301307 | ||
FQ227943.1 | - | 5 | 514 |
