siRNA Details
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199117.2 | NP_954548.1 | 4 | 410 | ||
XM_006242020.2 | XP_006242082.1 | 4 | 540 | |||
XM_006242021.2 | XP_006242083.1 | 4 | 540 | |||
XM_006242022.2 | XP_006242084.1 | 4 | 691 | |||
XM_006242024.2 | XP_006242086.1 | 4 | 523 | |||
XM_006242025.2 | XP_006242087.1 | 4 | 515 | |||
XM_006242026.2 | XP_006242088.1 | 4 | 524 | |||
GenBank mRNA | BC062392.1 | - | 4 | 390 | 5621139 |
