siRNA Details
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199117.2 | NP_954548.1 | 4 | 428 | ||
XM_006242020.2 | XP_006242082.1 | 4 | 558 | |||
XM_006242021.2 | XP_006242083.1 | 4 | 558 | |||
XM_006242022.2 | XP_006242084.1 | 4 | 709 | |||
XM_006242024.2 | XP_006242086.1 | 4 | 541 | |||
XM_006242025.2 | XP_006242087.1 | 4 | 533 | |||
XM_006242026.2 | XP_006242088.1 | 4 | 542 | |||
GenBank mRNA | BC062392.1 | - | 4 | 408 | 5621139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
