siRNA Details
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244855.1 | NP_001231784.1 | 2 | 303 | ||
XM_006242100.2 | XP_006242162.1 | 2 | 1743 | |||
XM_006242101.2 | XP_006242163.1 | 2 | 592 | |||
GenBank mRNA | BC082056.1 | - | 2 | 281 | 7132303 | |
CV126932.1 | - | 2 | 303 | |||
FQ217036.1 | - | 2 | 287 |

More Information
Other Species Targeted

Gene Ontology Categories:
