siRNA Details
Gene Symbol: Pnpla3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282324.1 | NP_001269253.1 | 2 | 307 | ||
GenBank mRNA | BC161881.1 | - | 2 | 625 |

More Information
Other Species Targeted

Gene Ontology Categories:
