siRNA Details
Gene Symbol: RGD1304694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271047.1 | NP_001257976.1 | 7 | 893 | ||
GenBank mRNA | BC098850.1 | - | 2 | 167 |

More Information
Other Species Targeted

