siRNA Details
Gene Symbol: RGD1306001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130696.1 | NP_001124168.1 | 2 | 180 | ||
XM_006242163.1 | XP_006242225.1 | 2 | 756 | |||
XM_008765765.1 | XP_008763987.1 | 2 | 344 |
