siRNA Details
Gene Symbol: Creld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037208.1 | NP_001032285.1 | 9 | 919 | ||
XM_006242216.2 | XP_006242278.1 | 8 | 774 | |||
GenBank mRNA | BC098722.1 | - | 9 | 919 | 7371287 | |
FQ229732.1 | - | 5 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
