siRNA Details
Gene Symbol: Siae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108759.1 | NP_001102229.1 | 2 | 135 | ||
GenBank mRNA | BC162050.1 | - | 2 | 106 |

More Information
Other Species Targeted

Gene Ontology Categories:
