siRNA Details
Gene Symbol: Cadm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012201.1 | NP_001012201.1 | 7 | 1067 | ||
XM_006242944.2 | XP_006243006.1 | 7 | 1066 | |||
XM_006242946.2 | XP_006243008.1 | 7 | 1066 | |||
XM_006242947.2 | XP_006243009.1 | 7 | 1066 | |||
XM_006242948.2 | XP_006243010.1 | 7 | 1066 | |||
GenBank mRNA | AB114443.1 | - | 7 | 956 | ||
AB257091.1 | - | 7 | 904 | |||
BC078966.1 | - | 7 | 1067 | 7111523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
