siRNA Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108761.1 | NP_001102231.1 | 6 | 947 | ||
XM_006243129.2 | XP_006243191.1 | 6 | 943 | |||
XM_006243131.2 | XP_006243193.1 | 5 | 730 | |||
XM_008766298.1 | XP_008764520.1 | 5 | 1135 | |||
XM_008766299.1 | XP_008764521.1 | 6 | 1218 | |||
XM_008766300.1 | XP_008764522.1 | 6 | 727 | |||
XM_008766301.1 | XP_008764523.1 | 5 | 661 | |||
XM_008766302.1 | XP_008764524.1 | 5 | 1529 |
