siRNA Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108761.1 | NP_001102231.1 | 8, 9 | 1564 | ||
XM_006243129.2 | XP_006243191.1 | 8, 9 | 1560 | |||
XM_006243131.2 | XP_006243193.1 | 7, 8 | 1347 | |||
XM_008766298.1 | XP_008764520.1 | 7, 8 | 1752 | |||
XM_008766299.1 | XP_008764521.1 | 8, 9 | 1835 | |||
XM_008766300.1 | XP_008764522.1 | 8, 9 | 1344 | |||
XM_008766301.1 | XP_008764523.1 | 7, 8 | 1278 | |||
XM_008766302.1 | XP_008764524.1 | 7, 8 | 2146 |
