siRNA Details
Gene Symbol: Fam219b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108764.1 | NP_001102234.1 | 2, 3 | 382 | ||
XM_008766309.1 | XP_008764531.1 | 2, 3 | 1582 | |||
XM_008766310.1 | XP_008764532.1 | 2, 3 | 1583 |

More Information
Other Species Targeted

