siRNA Details
Gene Symbol: Tipin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025287.1 | NP_001020458.1 | 6 | 673 | ||
GenBank mRNA | BC097351.1 | - | 6 | 673 | 7387433 |
Gene Symbol: Tipinl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730066.4 | XP_002730112.1 | 1 | 714 |
