siRNA Details
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008380.1 | NP_001008381.1 | 16, 17 | 2856 | ||
XM_006243261.2 | XP_006243323.1 | 16, 17 | 2959 | |||
XM_006243262.2 | XP_006243324.1 | 16, 17 | 2763 | |||
XM_006243263.2 | XP_006243325.1 | 16, 17 | 3288 | |||
GenBank mRNA | BC085932.1 | - | 16, 17 | 2856 | 7112012 | |
FQ216927.1 | - | 6, 7 | 1269 |
