siRNA Details
Gene Symbol: Ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009470.1 | NP_001009470.1 | 3 | 300 | ||
GenBank mRNA | BC088212.1 | - | 3 | 300 | 7383459 | |
BC097952.1 | - | 3 | 324 | 7454811 | ||
FQ228772.1 | - | 3 | 314 | |||
FQ234119.1 | - | 3 | 314 |
Gene Symbol: LOC100364016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758651.1 | XP_008756873.1 | 1 | 334 |
