siRNA Details
Gene Symbol: Slc17a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009713.2 | NP_001009713.1 | 5 | 704 | ||
GenBank mRNA | AY800277.1 | - | 5 | 703 | ||
BC097482.1 | - | 5 | 704 | 7459136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
