siRNA Details
Gene Symbol: Rwdd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108773.1 | NP_001102243.1 | 3 | 990 | ||
XM_006243483.2 | XP_006243545.1 | 3 | 877 | |||
XM_006243485.2 | XP_006243547.1 | 3 | 916 | |||
XM_006243486.2 | XP_006243548.1 | 3 | 1093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
