siRNA Details
Gene Symbol: Nmnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013224.1 | NP_001013242.1 | 2, 3 | 262 | ||
XM_006243608.2 | XP_006243670.1 | 2, 3 | 592 | |||
XM_006243609.2 | XP_006243671.1 | 2, 3 | 596 | |||
XM_006243610.2 | XP_006243672.1 | 2, 3 | 212 | |||
GenBank mRNA | BC083725.1 | - | 2, 3 | 262 | 7191168 |
