siRNA Details
Gene Symbol: Nmnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013224.1 | NP_001013242.1 | 3 | 290 | ||
XM_006243608.2 | XP_006243670.1 | 3 | 620 | |||
XM_006243609.2 | XP_006243671.1 | 3 | 624 | |||
XM_006243610.2 | XP_006243672.1 | 3 | 240 | |||
GenBank mRNA | BC083725.1 | - | 3 | 290 | 7191168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
