siRNA Details
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134520.1 | NP_001127992.1 | 16 | 2702 |
Gene Symbol: LOC102552166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766604.1 | XP_008764826.1 | 15 | 2932 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030730, AK155151, BC133714, AK083903, AK143506, and others.
Gene Ontology Categories:
