siRNA Details
Gene Symbol: Nprl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025744.2 | NP_001020915.2 | 2 | 288 | ||
GenBank mRNA | BC098932.1 | - | 2 | 267 | ||
CV077924.1 | - | 2 | 288 | |||
FQ209669.1 | - | 2 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
