siRNA Details
Gene Symbol: Dalrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014214.1 | NP_001014236.1 | 9 | 1238 | ||
GenBank mRNA | BC082046.1 | - | 9 | 1238 | 7131735 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
