siRNA Details
Gene Symbol: Elp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108782.2 | NP_001102252.1 | 7 | 751 | ||
GenBank mRNA | BC166773.1 | - | 7 | 751 | 7934150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
