siRNA Details
Gene Symbol: Crtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243992.2 | XP_006244054.1 | 3, 4 | 881 |
Gene Symbol: LOC102555086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244022.2 | XP_006244084.1 | 879 |

More Information
Other Species Targeted

Gene Ontology Categories:
