siRNA Details
Gene Symbol: Lyzl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001246183.1 | NP_001233112.1 | 3 | 310 | ||
XM_006244123.2 | XP_006244185.2 | 4 | 1553 | |||
XM_006244125.2 | XP_006244187.1 | 4 | 506 | |||
GenBank mRNA | HM125534.1 | - | 3 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
