siRNA Details
Gene Symbol: Zfp167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170577.1 | NP_001164048.1 | 6 | 1623 | ||
XM_006244197.2 | XP_006244259.2 | 6 | 2261 | |||
XM_006244199.1 | XP_006244261.1 | 6 | 1456 | |||
XM_006244200.2 | XP_006244262.1 | 6 | 1431 |
