siRNA Details
Gene Symbol: Sgol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766787.1 | XP_008765009.1 | 7 | 1258 | ||
XM_008766788.1 | XP_008765010.1 | 7 | 1227 | |||
XM_008766789.1 | XP_008765011.1 | 7 | 1255 |

More Information
Other Species Targeted

Gene Ontology Categories:
