siRNA Details
Gene Symbol: Sgol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766787.1 | XP_008765009.1 | 7 | 1251 | ||
XM_008766788.1 | XP_008765010.1 | 7 | 1220 | |||
XM_008766789.1 | XP_008765011.1 | 7 | 1248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
