siRNA Details
Gene Symbol: Tmem14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108790.1 | NP_001102260.1 | 449 | |||
XM_008766934.1 | XP_008765156.1 | 449 | ||||
GenBank mRNA | FQ213172.1 | - | 304 |
Gene Symbol: LOC103693210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766939.1 | XP_008765161.1 | 304 |

More Information
Other Species Targeted

