siRNA Details
Gene Symbol: Ccdc115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108793.1 | NP_001102263.1 | 3 | 696 | ||
GenBank mRNA | BC166583.1 | - | 261 | 7442489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
